3-Methyl-4-nitrobenzoic acid 99%

3-Methyl-4-nitrobenzoic acid

3-Methyl-4-nitrobenzoic acid 99%
Synonyms:
4-Nitro-m-toluic acid
Linear Formula:
CH3C6H3(NO2)CO2H
CAS Number:
Molecular Weight:
181.15
EC Number:

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
3-Methyl-4-nitrobenzoic acid, 99%