MilliporeSigma
3-Picoline analytical standard

3-Picoline

3-Picoline analytical standard
Synonyms:
3-Picoline, 3-Methylpyridine
Empirical Formula (Hill Notation):
C6H7N
CAS Number:
Molecular Weight:
93.13
EC Number:

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
3-Picoline
3-Picoline, analytical standard