MilliporeSigma
β-Sitosterol from soybean, ≥96%

β-Sitosterol

β-Sitosterol from soybean, ≥96%
Synonym(s):
α-Dihydrofucosterol, 22,23-Dihydrostigmasterol, 24α-Ethylcholesterol, 5-Stigmasten-3β-ol
Empirical Formula (Hill Notation):
C29H50O
CAS Number:
Molecular Weight:
414.71
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing