5-Methyl-2-hepten-4-one, predominantly trans 98%, FG

5-Methyl-2-hepten-4-one, predominantly trans

5-Methyl-2-hepten-4-one, predominantly trans 98%, FG
Synonyms:
Hazelnut ketone
Linear Formula:
C2H5CH(CH3)COCH=CHCH3
CAS Number:
Molecular Weight:
126.20

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort By: Default
Product Number
Product Description
Pricing
5-Methyl-2-hepten-4-one, predominantly trans, 98%, FG

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.