MilliporeSigma
Bifido Selective Supplement B suitable for microbiology

Acetic acid

Bifido Selective Supplement B suitable for microbiology
Synonym(s):
Glacial acetic acid, Acetic acid, Acetic acid solution
Linear Formula:
CH3CO2H
CAS Number:
Molecular Weight:
60.05
EC Number:

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique
Showing 1-20 of 23
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2