MilliporeSigma
1-Butanol for analysis EMPARTA® ACS

1-Butanol

1-Butanol for analysis EMPARTA® ACS
Synonyms:
n-Butanol, Butyl alcohol, 1-Butanol, Butyl Alcohol
Linear Formula:
CH3(CH2)3OH
CAS Number:
Molecular Weight:
74.12

Products

Shipping
Product Category
Brand
Sustainability
Boiling Point (°C)
Color
Feature
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
Technique