7-Methoxy-2-tetralone 97%

7-Methoxy-2-tetralone

7-Methoxy-2-tetralone 97%
Empirical Formula (Hill Notation):
C11H12O2
CAS Number:
Molecular Weight:
176.21
EC Number:

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
7-Methoxy-2-tetralone, 97%