MilliporeSigma
Paclitaxel, Semi-Synthetic A semi-synthetic form of Paclitaxel having no contamination by natural taxanes. Also has reduced levels of epimerization products as contaminants.

Paclitaxel, Taxus sp.

Paclitaxel, Semi-Synthetic A semi-synthetic form of Paclitaxel having no contamination by natural taxanes. Also has reduced levels of epimerization products as contaminants.
Synonym(s):
Paclitaxel, Taxus sp., Paclitaxel, Semi-Synthetic
Empirical Formula (Hill Notation):
C47H51NO14
CAS Number:
Molecular Weight:
853.91

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Research Area
Quality Segment
Special Grade