MilliporeSigma
Magnesium chloride AnhydroBeads™, −10 mesh, 99.99% trace metals basis

Magnesium chloride

Magnesium chloride AnhydroBeads™, −10 mesh, 99.99% trace metals basis
Synonym(s):
Magnogene, Magnesium chloride
Linear Formula:
MgCl2
CAS Number:
Molecular Weight:
95.21
EC Number:

Products

Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Particle Size (µm)
Special Grade
Technique