MilliporeSigma
ortho-Phosphoric acid 99% cryst. for analysis EMSURE®

Phosphoric acid

ortho-Phosphoric acid 99% cryst. for analysis EMSURE®
Synonym(s):
Orthophosphoric acid, Phosphoric acid
Linear Formula:
H3PO4
CAS Number:
Molecular Weight:
98.00

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Special Grade