ACA - CAS 110683-10-8 - Calbiochem A cell-permeable inhibitor of phospholipase A2 that blocks epinephrine-stimulated thromboxane production in platelets.

ACA - CAS 110683-10-8 - Calbiochem

ACA - CAS 110683-10-8 - Calbiochem A cell-permeable inhibitor of phospholipase A2 that blocks epinephrine-stimulated thromboxane production in platelets.
Synonyms:
ACA - CAS 110683-10-8 - Calbiochem
Empirical Formula (Hill Notation):
C21H23NO3
CAS Number:
Molecular Weight:
337.41

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort By: Default
Product Number
Product Description
Pricing
ACA - CAS 110683-10-8 - Calbiochem, A cell-permeable inhibitor of phospholipase A2 that blocks epinephrine-stimulated thromboxane production in platelets.

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.