Acetaminophen-(ring-d4) analytical standard

Acetaminophen-(ring-d4)

Acetaminophen-(ring-d4) analytical standard
Synonyms:
Paracetamol-(ring-d4), [2H4]-Paracetamol, [2H4]-Acetaminophen, [2H4]-Acetoaminophenol, N-(4-Hydroxyphenyl-2,3,5,6-d4)-acetamide, Acetaminophen-d4
Empirical Formula (Hill Notation):
C8D4H5NO2
CAS Number:
Molecular Weight:
155.19

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort By: Default
Product Number
Product Description
Pricing
Acetaminophen-(ring-d4), analytical standard

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.