Acetone-2,4-dinitrophenylhydrazone BCR®, certified reference material

Acetone-2,4-dinitrophenylhydrazone

Acetone-2,4-dinitrophenylhydrazone BCR®, certified reference material
Linear Formula:
(CH3)2C=NNHC6H3(NO2)2
CAS Number:
Molecular Weight:
238.20

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort By: Default
Product Number
Product Description
Pricing
Acetone-2,4-dinitrophenylhydrazone, environmental standard, 99%
Acetone-2,4-dinitrophenylhydrazone, BCR®, certified reference material

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.