AG 1295 - CAS 71897-07-9 - Calbiochem A cell-permeable, reversible, ATP-competitive, and selective inhibitor of platelet-derived growth factor (PDGF) receptor kinase (IC₅₀ = 500 nM) and PDGF-dependent DNA synthesis (IC₅₀ = 2.5 µM) in Swiss/3T3 cells.

AG 1295 - CAS 71897-07-9 - Calbiochem

AG 1295 - CAS 71897-07-9 - Calbiochem A cell-permeable, reversible, ATP-competitive, and selective inhibitor of platelet-derived growth factor (PDGF) receptor kinase (IC₅₀ = 500 nM) and PDGF-dependent DNA synthesis (IC₅₀ = 2.5 µM) in Swiss/3T3 cells.
Synonyms:
AG 1295 - CAS 71897-07-9 - Calbiochem
Empirical Formula (Hill Notation):
C16H14N2
CAS Number:
Molecular Weight:
234.30

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort By: Default
Product Number
Product Description
Pricing
AG 1295 - CAS 71897-07-9 - Calbiochem, A cell-permeable, reversible, ATP-competitive, and selective inhibitor of platelet-derived growth factor (PDGF) receptor kinase (IC₅₀ = 500 nM) and PDGF-dependent DNA synthesis (IC₅₀ = 2.5 µM) in Swiss/3T3 cells.

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.