AL-8810 ≥98% (HPLC), solid

AL-8810

AL-8810 ≥98% (HPLC), solid
Synonyms:
(5Z, 13E)-(9S,11S,15R)-9,15,Dihydroxy-11-fluoro-15-(2-indanyl)-16,17,18,19,20,pentanor-5,13-prostadienoic acid, [1R-[1α(Z),2β(1E,3S*),3β,5α]]-7-[3-Fluoro-5-hydroxy-2-[3-hydroxy-4-[2-(2,3-dihydro-1H-indenyl)]-1-butenyl]cyclolpentyl]-5-heptenoic acid
Empirical Formula (Hill Notation):
C24H31FO4
CAS Number:
Molecular Weight:
402.50

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort By: Default
Product Number
Product Description
Pricing
AL-8810, ≥98% (HPLC), solid

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.