ALLM - CAS 110115-07-6 - Calbiochem Cell permeable inhibitor of calpain I (Ki = 120 nM), calpain II (Ki = 230 nM), cathepsin B (Ki = 100 nM), and cathepsin L (Ki = 600 pM).

ALLM - CAS 110115-07-6 - Calbiochem

ALLM - CAS 110115-07-6 - Calbiochem Cell permeable inhibitor of calpain I (Ki = 120 nM), calpain II (Ki = 230 nM), cathepsin B (Ki = 100 nM), and cathepsin L (Ki = 600 pM).
Synonyms:
ALLM - CAS 110115-07-6 - Calbiochem
Empirical Formula (Hill Notation):
C19H35N3O4S
CAS Number:
Molecular Weight:
401.56

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort By: Default
Product Number
Product Description
Pricing
ALLM - CAS 110115-07-6 - Calbiochem, Cell permeable inhibitor of calpain I (Ki = 120 nM), calpain II (Ki = 230 nM), cathepsin B (Ki = 100 nM), and cathepsin L (Ki = 600 pM).

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.