Skip to Content
MilliporeSigma
Anti-ADNP antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Anti-ADNP antibody produced in rabbit

Anti-ADNP antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Synonym(s):
Anti-Activity-dependent neuroprotective protein antibody produced in rabbit, Anti-Activity-dependent neuroprotector antibody produced in rabbit, Anti-ADNP1, Anti-Activity-dependent neuroprotector homeobox, Anti-KIAA0784

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique