Skip to Content
MilliporeSigma
Anti-ALDH1A1 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Anti-ALDH1A1 antibody produced in rabbit

Anti-ALDH1A1 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Synonym(s):
Anti-ALDH-E1, Anti-ALHDII, Anti-Aldehyde dehydrogenase family 1 member A1, Anti-Aldehyde dehydrogenase, cytosolic, Anti-RALDH 1, Anti-RalDH1, Anti-Retinal dehydrogenase 1

Products

Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Application
Post Translational Modification
Species Reactivity
Technique