Skip to Content
MilliporeSigma
Anti-CD19 antibody, Rabbit monoclonal recombinant, expressed in HEK 293 cells, clone RM332, purified immunoglobulin

Anti-CD19 antibody, Rabbit monoclonal

Anti-CD19 antibody, Rabbit monoclonal recombinant, expressed in HEK 293 cells, clone RM332, purified immunoglobulin

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Recombinant Host
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique