
Anti-c-Fos (Ab-2) (4-17) Rabbit pAb

Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Quality Segment
Post Translational Modification
Species Reactivity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing