Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Anti-ERICH4 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Anti-ERICH4 antibody produced in rabbit

Anti-ERICH4 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique