Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now
Anti-gp120 antibody produced in goat affinity isolated antibody

Anti-gp120 antibody produced in goat

Anti-gp120 antibody produced in goat affinity isolated antibody
Synonym(s):
Anti-HIV-1 glycoprotein 120

Products

Product Category
Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing