Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day through March 27.Save Now
Anti-INSL3 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Anti-INSL3 antibody produced in rabbit

Anti-INSL3 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Synonym(s):
Anti-MGC119818, Anti-MGC119819, Anti-RLF, Anti-RLNL, Anti-insulin-like 3 (Leydig cell)

Products

Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique