Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Anti-KIF11 antibody produced in rabbit affinity isolated antibody, Prestige Antibodies® Powered by Atlas Antibodies, buffered aqueous glycerol solution

Anti-KIF11 antibody produced in rabbit

Anti-KIF11 antibody produced in rabbit affinity isolated antibody, Prestige Antibodies® Powered by Atlas Antibodies, buffered aqueous glycerol solution
Synonym(s):
Anti-Kinesin-like protein 1, Anti-Kinesin-like protein KIF11, Anti-Kinesin-like spindle protein HKSP, Anti-Kinesin-related motor protein Eg5, Anti-TRIP-5, Anti-Thyroid receptor-interacting protein 5, Eg5, KIFL1, KNSL1, Kinesin-like protein 1, Kinesin-like protein KIF11, HKSP, TRIP5

Products

Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique