Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now
Anti-NCF1 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Anti-NCF1 antibody produced in rabbit

Anti-NCF1 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Synonym(s):
Anti-NCF1A, Anti-NOXO2, Anti-SH3PXD1A, Anti-p47phox

Products

Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique