Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now
Anti-SERPINA3 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Anti-SERPINA3 antibody produced in rabbit

Anti-SERPINA3 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Synonym(s):
Anti-α-1-antichymotrypsin, Anti-Aact, Anti-Act, Anti-Serpin peptidase inhibitor, clade a (α-1 antiproteinase, antitrypsin), member 3, Anti-ACT, Anti-Alpha-1-antichymotrypsin precursor, Anti-Cell growth-inhibiting gene 24/25 protein

Products

Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique