
Anti-SLC25A22 antibody produced in rabbit

Synonym(s):
Anti-FLJ13044, Anti-GC1, Anti-Solute carrier family 25 (mitochondrial carrier: glutamate), member 22
Products
Product Category
Brand
Biological Source
Clonality
Physical Form
Product Line
Research Area
Conjugate
Quality Segment
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing