Skip to Content
MilliporeSigma
Anti-SOST (center) antibody produced in rabbit Ig fraction of antiserum, buffered aqueous solution

ANTI-SOST (CENTER) antibody produced in rabbit

Anti-SOST (center) antibody produced in rabbit Ig fraction of antiserum, buffered aqueous solution
Synonym(s):
SOST, Sclerostin

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique