Skip to Content
MilliporeSigma
Anti-Sox9 Antibody Chemicon®, from rabbit

Anti-SOX9 Antibody

Anti-Sox9 Antibody Chemicon®, from rabbit
Synonym(s):
Anti-Transcription factor SOX-9 antibody produced in rabbit, CMD1, CMPD1, SRA1, SRY sex determining region Y-box 9 protein, SRY-box containing gene 9, Sox-9, transcription factor SOX-9, Anti-CMD1, Anti-CMPD1, Anti-SRA1, Anti-SRY (sex determining region Y)-box 9 (campomelic dysplasia, autosomal sex-reversal)

Products

Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Isotype
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique