Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day through March 27.Save Now
Anti-TAP1 antibody produced in rabbit affinity isolated antibody

Anti-TAP1 antibody produced in rabbit

Anti-TAP1 antibody produced in rabbit affinity isolated antibody
Synonym(s):
Anti-ABC17, Anti-ABCB2, Anti-APT1, Anti-D6S114E, Anti-Transporter 1, ATP-binding cassette, sub-family B (MDR/TAP)

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique