Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now
Anti-Tryptophan Hydroxylase Antibody Chemicon®, from sheep

Anti-Tryptophan Hydroxylase Antibody

Anti-Tryptophan Hydroxylase Antibody Chemicon®, from sheep
Synonym(s):
Anti-TPH, Anti-Tryptophan 5-hydroxylase 1, Anti-Tryptophan 5-monooxygenase 1, TPH, TRPH, tryptophan 5-hydroxylase 1, tryptophan 5-monooxygenase 1

Products

Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Isotype
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique