Skip to Content
MilliporeSigma
Anti-ZMYND8 antibody produced in rabbit

Anti-ZMYND8 antibody produced in rabbit

Anti-ZMYND8 antibody produced in rabbit
Synonym(s):
Anti-CTCL tumor antigen se14-3, Anti-Cutaneous T-cell lymphoma-associated antigen se14-3, Anti-Protein kinase C-binding protein 1, Anti-Rack7, Anti-Zinc finger MYND domain-containing protein 8

Products

Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique