Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Anti-ZNF26 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Anti-ZNF26 antibody produced in rabbit

Anti-ZNF26 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Synonym(s):
Anti-Zinc finger protein 26, Anti-Zinc finger protein KOX20

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique