MilliporeSigma
Ascentis® Express Biphenyl Column 2.7 μm particle size, L × I.D. 7.5 cm × 4.6 mm

Ascentis® Express Biphenyl Column

Ascentis® Express Biphenyl Column 2.7 μm particle size, L × I.D. 7.5 cm × 4.6 mm

Products

Product Category
Brand
pH Value
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Quality Segment
Application
Package Size
Particle Size (µm)
Pore Size (Å)
Technique

Products

Showing 1-16 of 16
Compare
Product Number
Description
Pricing