Skip to Content
Products
Cart
0
US
EN
Products
Products
Applications
Services
Resources
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Lab Automation
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Documents
Learning Center
New
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
0
Back
4-(4-Methoxyphenyl)-4-oxobut-2-enoic acid
CAS Number:
5711-41-1
Molecular Weight:
206.2
Products
Brand
Ambeed, Inc. (1)
ChemScene LLC (Preferred Partner) (1)
Purity
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
CIAH99F8E876
4-(4-Methoxyphenyl)-4-oxobut-2-enoic acid,
Expand
View Pricing
AMBH9614F479
4-(4-Methoxyphenyl)-4-oxobut-2-enoic acid,
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
English - EN
Español - ES
Learn More