Skip to Content
US
EN
Products
Applications
Services
Resources
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Lab Automation
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Documents
Learning Center
New
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
4-Butylbenzoic acid
CAS Number:
20651-71-2
Molecular Weight:
178.23
Products
Brand
AiFChem (an XtalPi company)
Aladdin Scientific
ChemScene LLC (Preferred Partner)
Synthonix Corporation
Purity
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
ALNH9A9DB53F
4-Butylbenzoic acid,
Expand
View Pricing
SY3H3D67892B
4-Butylbenzoic acid,
Expand
View Pricing
XPIH9BD066FF
4-Butylbenzoic acid,
Expand
View Pricing
CIAH987F6400
4-Butylbenzoic acid,
Expand
View Pricing