Skip to Content
US
EN
Products
Applications
Services
Resources
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Lab Automation
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Documents
Learning Center
New
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Get
22% off
for Pi Day through March 31.
Save Now
Back
5-Iodopent-1-yne
CAS Number:
2468-55-5
Molecular Weight:
194.02
Products
Brand
AiFChem (an XtalPi company)
ChemScene LLC (Preferred Partner)
Purity
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
XPIH9BD016E8
5-Iodopent-1-yne,
Expand
View Pricing
CIAH987F1DB9
5-Iodopent-1-yne,
Expand
View Pricing