Skip to Content
US
EN
Products
Applications
Services
Resources
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Lab Automation
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Documents
Learning Center
New
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Chlorogenic Acid
CAS Number:
327-97-9
Molecular Weight:
354.31
Products
Brand
Apollo Scientific Ltd (1)
Chem Impex International, Inc. (1)
Targetmol Chemicals Inc. (1)
Purity
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
TA9H9A175F10
Chlorogenic Acid,
Expand
View Pricing
APOH11A9CD84
Chlorogenic Acid,
Expand
View Pricing
CH6H9A56D543
Chlorogenic Acid,
Expand
View Pricing