Skip to Content
MilliporeSigma
Linolenic acid, 463-40-1, undefined,

Linolenic acid

Linolenic acid, 463-40-1, undefined,
CAS Number:
Molecular Weight:
278.44

Products

Brand
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing