Benazolin-ethyl ester PESTANAL®, analytical standard

Benazolin-ethyl ester

Benazolin-ethyl ester PESTANAL®, analytical standard
Synonyms:
Ethyl-4-chlor-2-oxo-benzothiazolinacetat, 4-Chloro-2-oxo-3(2H)-benzothiazoleacetic acid ethyl ester
Empirical Formula (Hill Notation):
C11H10ClNO3S
CAS Number:
Molecular Weight:
271.72
EC Number:

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort By: Default
Product Number
Product Description
Pricing
Benazolin-ethyl ester, PESTANAL®, analytical standard

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.