Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now
β-Ionone for synthesis

β-Ionone

β-Ionone for synthesis
Synonym(s):
4-(2,6,6-Trimethyl-1-cyclohexenyl)-3-buten-2-one, β-Ionone
Empirical Formula (Hill Notation):
C13H20O
CAS Number:
Molecular Weight:
192.30

Products

Shipping
Product Category
Brand
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing