Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Bismuth(III) oxide purum, ≥98.0% (KT)

Bismuth oxide

Bismuth(III) oxide purum, ≥98.0% (KT)
Synonym(s):
Dibismuth trioxide, Bismuth(III) oxide
Empirical Formula (Hill Notation):
Bi2O3
CAS Number:
Molecular Weight:
465.96
EC Number:

Products

Shipping
Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Particle Size (µm)
Reagent Type
Core
Special Grade