Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now
Blood typing Bulk Reagent - Rare Specificities & AHG Anti-S from the clone MS-94, Human IgM, Blood Grouping Reagent

Blood typing Bulk Reagent - Rare Specificities & AHG

Blood typing Bulk Reagent - Rare Specificities & AHG Anti-S from the clone MS-94, Human IgM, Blood Grouping Reagent

Products

Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Quality Segment
Package Size
Technique
Showing 1-14 of 14
Compare
Product Number
Description
Pricing