
Blood typing Intermediates for Further Manufacturing Use (FFMU) - ABO Specificities

Synonym(s):
Anti-A (clone BIRMA-1), FFMU
Products
Product Category
Brand
Biological Source
Clonality
Physical Form
Quality Segment
Technique
Showing 1-8 of 8
Compare
Product Number
Description
Pricing