MilliporeSigma
(Boc-aminooxy)acetic acid ≥98.0% (T)

(Boc-aminooxy)acetic acid

(Boc-aminooxy)acetic acid ≥98.0% (T)
Synonym(s):
N-Boc-(carboxymethoxy)amine, Boc-amino-oxyacetic acid
Linear Formula:
(CH3)3CO2CNHOCH2CO2H
CAS Number:
Molecular Weight:
191.18

Products

Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Reagent Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing