Boc-D-Phg-OH ≥99.0% (TLC)

Boc-D-Phg-OH

Boc-D-Phg-OH ≥99.0% (TLC)
Synonyms:
Boc-D-Phg-OH, Boc-D-α-phenylglycine
Linear Formula:
C6H5CH(COOH)NHCOOC(CH3)3
CAS Number:
Molecular Weight:
251.28

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort By: Default
Product Number
Product Description
Pricing
Boc-D-Phg-OH, Novabiochem®
Boc-D-Phg-OH, ≥99.0% (TLC)