US
EN
Applications
Products
Services
Support
Home
Search Results
Boc-Glu(OcHx)-OH
(1)
Boc-Glu(OcHx)-OH
Synonyms:
Boc-Glu(OcHx)-OH
Empirical Formula (Hill Notation):
C
16
H
27
NO
6
CAS Number:
73821-97-3
Molecular Weight:
329.39
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
8.53029
Boc-Glu(OcHx)-OH, Novabiochem
®
Expand
View Pricing