Skip to Content
MilliporeSigma
Boc-His(Trt)-OH ≥98.0% (TLC)

Boc-His(Trt)-OH

Boc-His(Trt)-OH ≥98.0% (TLC)
Synonym(s):
Nα-Boc-N(im)-trityl-L-histidine, Boc-His(Trt)-OH
Empirical Formula (Hill Notation):
C30H31N3O4
CAS Number:
Molecular Weight:
497.58

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing