Skip to Content
MilliporeSigma
Boc-Tyr(tBu)-OH ≥99.0% (sum of enantiomers, TLC)

Boc-Tyr(tBu)-OH

Boc-Tyr(tBu)-OH ≥99.0% (sum of enantiomers, TLC)
Synonym(s):
Boc-Tyr(tBu)-OH, Boc-O-tert-butyl-L-tyrosine
Linear Formula:
(CH3)3COC6H4CH2CH(COOH)NHCOOC(CH3)3
CAS Number:
Molecular Weight:
337.41

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing