Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Boron Cell Test photometric, 0.05-2.00 mg/L (B), Spectroquant®

Boron Test, photometric

Boron Cell Test photometric, 0.05-2.00 mg/L (B), Spectroquant®

Products

Product Category
Brand
Product Line
Analyte Suitability
Compatibility
Detection Method
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing